EMAIL THIS PAGE TO A FRIEND
P6385 Supelco

Protoveratrine A

≥80%

Purchase

Properties

InChI Key   HYTGGNIMZXFORS-MGYKWWNKSA-N
assay   ≥80%
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1544PSN1 6.1 / PGI
WGK Germany 
3
RTECS 
FL5750100

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?