• USA Home
  • P6535 - D-Phe-Met-Arg-Phe-NH2

EMAIL THIS PAGE TO A FRIEND
P6535 Sigma-Aldrich

D-Phe-Met-Arg-Phe-NH2

≥97% (HPLC)

Synonym: [D-Phe1]-Molluscan cardioexcitatory neuropeptide

  • CAS Number 84313-42-8

  • Empirical Formula (Hill Notation) C29H42N8O4S

  • Molecular Weight 598.76

  •  MDL number MFCD00076626

Purchase

Properties

assay   ≥97% (HPLC)
composition   Peptide content, ~65%
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?