• USA Home
  • P6906 - Phenoxyacetic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
P6906 Sigma-Aldrich

Phenoxyacetic acid sodium salt

Purchase

Properties

SMILES string   [Na+].[O-]C(=O)COc1ccccc1
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AJ2400000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?