• USA Home
  • P6910 - Phe-Met-D-Arg-Phe amide

EMAIL THIS PAGE TO A FRIEND
P6910 Sigma-Aldrich

Phe-Met-D-Arg-Phe amide

≥97% (HPLC)

Synonym: [D-Arg3]-Molluscan cardioexcitatory neuropeptide

  • CAS Number 84313-44-0

  • Empirical Formula (Hill Notation) C29H42N8O4S

  • Molecular Weight 598.76

  •  MDL number MFCD00076628

Purchase

Properties

assay   ≥97% (HPLC)
composition   Peptide content, ~70%
storage temp.   −20°C
SMILES string   CSCCC(NC(=O)C(N)Cc1ccccc1)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc2ccccc2)C(N)=O
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?