• USA Home
  • P6967 - Penta-N-acetylchitopentaose

EMAIL THIS PAGE TO A FRIEND
P6967 Sigma-Aldrich

Penta-N-acetylchitopentaose

≥95%

Synonym: NI, NII, NIII, NIV, NV-Pentaacetylchitopentaose

  • CAS Number 36467-68-2

  • Empirical Formula (Hill Notation) C40H67N5O26

  • Molecular Weight 1033.98

  •  MDL number MFCD00210240

Purchase

Properties

assay   ≥95%
storage temp.   −20°C

Description

Substrates

Elicits plant defense systems; carbohydrate component of lipo-chitooligosaccharides secreted by Rhizobia, functioning as a signaling molecule in the host plant; substrate for Rhizobium leguminosarum nodulation protein NodL.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?