• USA Home
  • P7152 - Prochlorperazine edisylate salt

EMAIL THIS PAGE TO A FRIEND
P7152 Sigma-Aldrich

Prochlorperazine edisylate salt

  • CAS Number 1257-78-9

  • Empirical Formula (Hill Notation) C20H24ClN3S · C2H6O6S2

  • Molecular Weight 564.14

  •  EC Number 215-019-1

  •  MDL number MFCD00056797

Purchase

Properties

storage temp.   2-8°C

Description

Biochem/physiol Actions

Phenothiazine antipsychotic; D2 antagonist; antispasmodic

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
SO2975000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?