• USA Home
  • P7549 - Pyrantel citrate salt

EMAIL THIS PAGE TO A FRIEND
P7549 Sigma-Aldrich

Pyrantel citrate salt

Synonym: 1-Methyl-2-(2-[2-thienyl]ethenyl)-1,4,5,6-tetrahydropyrimidine

Purchase

Properties

Mode of action   enzyme | inhibits
antibiotic activity spectrum   parasites
SMILES string   OC(=O)CC(O)(CC(O)=O)C(O)=O.CN1CCCN=C1\C=C\c2cccs2
InChI   1S/C11H14N2S.C6H8O7/c1-13-8-3-7-12-11(13)6-5-10-4-2-9-14-10;7-3(8)1-6(13,5(11)12)2-4(9)10/h2,4-6,9H,3,7-8H2,1H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/b6-5+;
InChI key   YJGGCARNRYGSPA-IPZCTEOASA-N

Description

Application

Pyrantel is an antinematodal thiophene. It is commonly used by veterinarians to treat and prevent intestinal parasites in small animals. It has been used to phenotypically characterize Ancylostoma caninum isolates and is used to distinguish cholinergic receptor subtypes in Ascaris muscle.

Biochem/physiol Actions

Pyrantel is a nematode muscle cell-associated nicotinic agonist for the acetylcholine (ACh) receptor. Cholinesterase inhibitor.

Pyrantel is a nematode muscle cell-associated nicotinic agonist for the acetylcholine (ACh) receptor. This is a cholinesterase inhibitor. It causes muscle paralysis in parasitic worms due to prolonged activation of the excitatory nicotinic acetylcholine (nACh) receptors.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Storage class (TRGS 510): Non Combustible Solids

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?