• USA Home
  • P7584 - (−)-Bis[(S)-1-phenylethyl]amine hydrochloride

EMAIL THIS PAGE TO A FRIEND
P7584 Sigma-Aldrich

(−)-Bis[(S)-1-phenylethyl]amine hydrochloride

≥98% (titration)

  • CAS Number 40648-92-8

  • Empirical Formula (Hill Notation) C16H19N · HCl

  • Molecular Weight 261.79

  •  MDL number MFCD00216671

Purchase

Properties

assay   ≥98% (titration)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?