• USA Home
  • P7674 - Pyrantel (+)-tartrate salt

EMAIL THIS PAGE TO A FRIEND
P7674 Sigma-Aldrich

Pyrantel (+)-tartrate salt

Synonym: 1-Methyl-2-(2-[2-thienyl]ethenyl)-1,4,5,6-tetrahydropyrimidine

Purchase

Properties

Related Categories Antibiotics, Antibiotics A to Z, Antibiotics N-S, Antiparasitic / Antiprotozoal / Anthelminthic, Biochemicals and Reagents,
solubility   H2O: 4.0 mL, clear, colorless to faint yellow or tan (Solvent: 200 mg plus 4.0 mL )
Mode of action   enzyme | inhibits
antibiotic activity spectrum   parasites
SMILES string   O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCCN=C1\C=C\c2cccs2
InChI   1S/C11H14N2S.C4H6O6/c1-13-8-3-7-12-11(13)6-5-10-4-2-9-14-10;5-1(3(7)8)2(6)4(9)10/h2,4-6,9H,3,7-8H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/b6-5+;/t;1-,2-/m.1/s1
InChI key   VWRCYAZJKNPEQR-NIEARKAZSA-N

Description

Application

Pyrantel is used to study neuronal nicotinic receptors and has been is used to treat parasiticinfections in livestock. Pyrantel tartrate salt is used to study allosteric modulation of the neuronal nicotinic acetylcholine receptors (nAChRs).

Biochem/physiol Actions

Pyrantel is a nematode muscle cell-associated nicotinic agonist for the acetylcholine (ACh) receptor. Cholinesterase inhibitor.

Pyrantel is a nematode muscle cell-associated nicotinic agonist for the acetylcholine (ACh) receptor. This is a cholinesterase inhibitor. It causes spastic muscle paralysis in parasitic worms due to prolonged activation of the excitatory nicotinic acetylcholine (nACh) receptors.

Other Notes

Keep container tightly closed in a dry and well-ventilated place

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
UW0249000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?