EMAIL THIS PAGE TO A FRIEND
P7744 Sigma-Aldrich

Potassium iodide

meets EP, BP, USP testing specifications

Purchase

Properties

Agency/Method   meets BP testing specifications
  meets EP testing specifications
  meets USP testing specifications
vapor pressure   1 mmHg ( 745 °C)
quality   meets EP, BP, USP testing specifications
mp   681 °C (lit.)
SMILES string   [K+].[I-]
InChI   1S/HI.K/h1H;/q;+1/p-1
InChI key   NLKNQRATVPKPDG-UHFFFAOYSA-M
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TT2975000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?