• USA Home
  • P7902 - Phosphomycin calcium salt

EMAIL THIS PAGE TO A FRIEND
P7902 Sigma-Aldrich

Phosphomycin calcium salt

Synonym: (−)-(1R,2S)-(1,2-Epoxypropyl)phosphonic acid, Fosfomycin calcium salt, Phosphonomycin

Purchase

Properties

Related Categories Antibacterial, Antibiotics, Antibiotics A to Z, Antibiotics N-S, Antibiotics by Mechanism of Action,
solubility   1 M HCl: soluble 50 mg/mL
Mode of action   cell wall synthesis | interferes
antibiotic activity spectrum   Gram-negative bacteria
  Gram-positive bacteria
storage temp.   2-8°C
SMILES string   [Ca++].CC1OC1P([O-])([O-])=O
InChI   1S/C3H7O4P.Ca/c1-2-3(7-2)8(4,5)6;/h2-3H,1H3,(H2,4,5,6);/q;+2/p-2
InChI key   YMZJBJPWTXJQMR-UHFFFAOYSA-L

Description

Application

Phosphomycin, also known as Fosfomycin, is an antibiotic produced by Streptomyces fradiae. It is used to treat uncomplicated urinary tract infections and to reduce nephrotoxicity and ototoxicity of platinum-containing anti-tumor agents. It is used to study resistance proteins in Bacillus subtilis and Mycobacterium tuberculosis. It is a potential treatment for bronchiectasis.

Packaging

1, 5, 10 g in poly bottle

Biochem/physiol Actions

Fosfomycin is a phosphoenolpyruvate analog. It irreversibly inhibits enolpyruvate transferase (MurA). Therefore, the producion of N-acetylmuramic acid, an essential element of the peptidoglycan cell wall is inhibited.

Phosphomycin calcium salt blocks the formation or N-acetylmuramic acid by inhibiting UDP-Nacetylglucosamine 3-enolpyruvate transferase (MurA).

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?