• USA Home
  • P8258 - 5β-Pregnane-3α,17α,20β-triol

EMAIL THIS PAGE TO A FRIEND
P8258 Sigma-Aldrich

5β-Pregnane-3α,17α,20β-triol

Synonym: 3α,17α,20β-Trihydroxy-5β-pregnane

  • CAS Number 1165-28-2

  • Empirical Formula (Hill Notation) C21H36O3

  • Molecular Weight 336.51

  •  EC Number 214-614-3

Purchase
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?