• USA Home
  • P8631 - 3α,17α-Dihydroxy-5β-pregnane-11,20-dione

EMAIL THIS PAGE TO A FRIEND
P8631 Sigma-Aldrich

3α,17α-Dihydroxy-5β-pregnane-11,20-dione

Synonym: 17α-Hydroxy-11-ketopregnanolone, 5β-Pregnane-3α,17α-diol-11,20-dione

  • CAS Number 641-78-1

  • Empirical Formula (Hill Notation) C21H32O4

  • Molecular Weight 348.48

  •  EC Number 211-376-2

Purchase

Properties

SMILES string   CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?