EMAIL THIS PAGE TO A FRIEND
P8911 Sigma-Aldrich

Pentacosanoic acid

≥99% (capillary GC)

Purchase

Properties

assay   ≥99% (capillary GC)
storage temp.   2-8°C
SMILES string   CCCCCCCCCCCCCCCCCCCCCCCCC(O)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?