• USA Home
  • P9055 - L-Phenylalanine-UL-14C hydrochloride

EMAIL THIS PAGE TO A FRIEND
P9055 Sigma-Aldrich

L-Phenylalanine-UL-14C hydrochloride

≥98% (Radiochemical Purity, HPLC), aqueous ethanol solution

Synonym: (S)-(−)-phenylalanine-UL-14C

  • CAS Number 658-69-5

  • Linear Formula 14C6H514CH214CH[NH2]14COOH

  • Molecular Weight 165.19

  •  MDL number MFCD00069585

  •  PubChem Substance ID 24899006

Purchase

Properties

assay   ≥98% (Radiochemical Purity, HPLC)
form   aqueous ethanol solution
extent of labeling   360-540 mCi per mmol
mass shift   M+6
storage temp.   2-8°C

Description

Packaging

Combi-vial

Physical form

Aqueous solution containing 2% ethanol

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?