• USA Home
  • P9079 - N2-Phenoxyacetyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyguanosine

EMAIL THIS PAGE TO A FRIEND
P9079 Sigma-Aldrich

N2-Phenoxyacetyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyguanosine

~98%

  • CAS Number 115388-95-9

  • Empirical Formula (Hill Notation) C39H37N5O8

  • Molecular Weight 703.74

  •  MDL number MFCD00079255

Purchase

Properties

assay   ~98%
storage temp.   −20°C
SMILES string   COc1ccc(cc1)C(OC2CC(OC2COC(=O)COc3ccccc3)n4cnc5C(=O)N=C(N)Nc45)(c6ccccc6)c7ccc(OC)cc7
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?