• USA Home
  • P9153 - DL-Pantothenic acid hemicalcium salt

EMAIL THIS PAGE TO A FRIEND
P9153 Sigma-Aldrich

DL-Pantothenic acid hemicalcium salt

≥97% (TLC)

Synonym: (±)-N-(2,4-Dihydroxy-3,3-dimethylbutyryl)-β-alanine, DL-N-(2,4-Dihydroxy-3,3-dimethylbutyryl)-β-alanine, Calcium pantothenate

  • CAS Number 63409-48-3

  • Linear Formula HOCH2C(CH3)2CH(OH)CONHCH2CH2CO2 · 1/2Ca2+

  • Molecular Weight 238.27

  •  MDL number MFCD00063189

  •  PubChem Substance ID 24899013

Purchase

Properties

Related Categories Biochemicals and Reagents, Vitamin B, Vitamin B5, Vitamins
InChI Key   FAPWYRCQGJNNSJ-UHFFFAOYSA-L
assay   ≥97% (TLC)
form   powder
color   white to light tan
mp   138 °C (dec.) (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?