• USA Home
  • P9157 - N-Palmitoyl-D-sphingosine semisynthetic from bovine brain D-sphingosine

EMAIL THIS PAGE TO A FRIEND
P9157 Sigma

N-Palmitoyl-D-sphingosine semisynthetic from bovine brain D-sphingosine

≥99% (TLC), powder

Synonym: C16 ceramide, N-Hexadecanoyl-D-sphingosine, N-Palmitoyl-D-erythro-sphingosine, Palmitoyl ceramide

  • CAS Number 24696-26-2

  • Empirical Formula (Hill Notation) C34H67NO3

  • Molecular Weight 537.90

  •  MDL number MFCD00056205

Purchase

Properties

assay   ≥99% (TLC)
form   powder
color   white
solubility   ethanol: soluble (warm)
storage temp.   −20°C

Description

Biochem/physiol Actions

Naturally occurring ceramide; induces EGF receptor phosphorylation in A-431 cells; activates phospholipase A2.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?