• USA Home
  • P9260 - N-Palmitoyl-DL-dihydrogalactocerebroside

EMAIL THIS PAGE TO A FRIEND
P9260 Sigma-Aldrich

N-Palmitoyl-DL-dihydrogalactocerebroside

Synonym: 1-O-(β-D-Galactopyranosyl)-N-hexadecanoyl-DL-dihydrosphingosine

Purchase

Properties

Related Categories Biochemicals and Reagents, Ceramides, Complex Lipids, Glycolipids, Lipids,
form   solid
shipped in   ambient
storage temp.   −20°C
SMILES string   CCCCCCCCCCCCCCCC(O)C(COC1OC(CO)C(O)C(O)C1O)NC(=O)CCCCCCCCCCCCCCC
InChI   1S/C40H79NO8/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(43)33(32-48-40-39(47)38(46)37(45)35(31-42)49-40)41-36(44)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33-35,37-40,42-43,45-47H,3-32H2,1-2H3,(H,41,44)
InChI key   BLGKYYVFGMKTEZ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?