EMAIL THIS PAGE TO A FRIEND
P9268 Sigma-Aldrich

Phorbol 12-myristate

  • CAS Number 20839-06-9

  • Empirical Formula (Hill Notation) C34H54O7

  • Molecular Weight 574.79

  •  MDL number MFCD00036782

Purchase

Properties

storage temp.   −20°C
SMILES string   CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@@]2(O)[C@@H](C=C(CO)C[C@@]3(O)[C@H]2C=C(C)C3=O)[C@@H]4C(C)(C)[C@]14O
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
XB8580000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?