EMAIL THIS PAGE TO A FRIEND
P9317 Sigma-Aldrich

Pneumadin Human

≥97% (HPLC)

Synonym: Ala-Gly-Glu-Pro-Lys-Leu-Asp-Ala-Gly-Val-NH2

  • CAS Number 130918-91-1

  • Empirical Formula (Hill Notation) C41H70N12O14

  • Molecular Weight 955.07

  •  MDL number MFCD00153585

Purchase

Properties

assay   ≥97% (HPLC)
composition   Peptide content, ~70%
storage temp.   −20°C
SMILES string   CC(C)CC(NC(=O)C(CCCCN)NC(=O)C1CCCN1C(=O)C(CCC(O)=O)NC(=O)CNC(=O)C(C)N)C(=O)NC(CC(O)=O)C(=O)NC(C)C(=O)NCC(=O)NC(C(C)C)C(N)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?