• USA Home
  • P9414 - 3-Phenyl-3,4-dihydroisocoumarin

EMAIL THIS PAGE TO A FRIEND
P9414 Sigma-Aldrich

3-Phenyl-3,4-dihydroisocoumarin

  • CAS Number 2674-44-4

  • Empirical Formula (Hill Notation) C15H12O2

  • Molecular Weight 224.25

  •  MDL number MFCD00030743

Purchase

Properties

SMILES string   O=C1OC(Cc2ccccc12)c3ccccc3
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?