• USA Home
  • P9525 - β-Acetyl-γ-O-alkyl-L-α-phosphatidylcholine from bovine heart lecithin

EMAIL THIS PAGE TO A FRIEND
P9525 Sigma-Aldrich

β-Acetyl-γ-O-alkyl-L-α-phosphatidylcholine from bovine heart lecithin

2 mg/mL in chloroform, ~99%

Synonym: 1-O-Alkyl-2-acetyl-sn-glycero-3-phosphocholine, 2-Acetyl-1-alkyl-sn-glycero-3-phosphocholine, AcGEPC, PAF, Platelet activating factor

Purchase

Properties

assay   ~99%
concentration   2 mg/mL in chloroform
storage temp.   −20°C
SMILES string   [O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(C)=O)COCCCCCCCCCCCCCCCC)=O
InChI   1S/C26H54NO7P/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-19-21-31-23-26(34-25(2)28)24-33-35(29,30)32-22-20-27(3,4)5/h26H,6-24H2,1-5H3/t26-/m1/s1
InChI key   HVAUUPRFYPCOCA-AREMUKBSSA-N

Description

Biochem/physiol Actions

Phospholipid produced by lymphocytes, platelets and endothelial cells that induces platelet aggregation and is involved in inflammation, anaphylaxis and wound repair.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 1888 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?