• USA Home
  • P9893 - 20-Deoxy-20-oxophorbol 12-myristate 13-acetate

EMAIL THIS PAGE TO A FRIEND
P9893 Sigma-Aldrich

20-Deoxy-20-oxophorbol 12-myristate 13-acetate

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C13-C60, Carbonyl Compounds, Chemical Synthesis,
storage temp.   −20°C
SMILES string   CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@@]2(O)C(C=C(C[C@@]3(O)C2C=C(C)C3=O)C=O)C4C(C)(C)[C@]14OC(C)=O
InChI   1S/C36H54O8/c1-7-8-9-10-11-12-13-14-15-16-17-18-29(39)43-32-24(3)35(42)27(30-33(5,6)36(30,32)44-25(4)38)20-26(22-37)21-34(41)28(35)19-23(2)31(34)40/h19-20,22,24,27-28,30,32,41-42H,7-18,21H2,1-6H3/t24-,27+,28-,30-,32-,34-,35-,36-/m1/s1
InChI key   AHLWZBVXSWOPPL-RGYGYFBISA-N

Description

Application

Reduction with tritiated NaBH4 yields tritiated phorbol 12-myristate 13-acetate that can be used for receptor binding studies

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?