• USA Home
  • PZ0145 - PD-160725 2-hydroxyethanesulfonate

EMAIL THIS PAGE TO A FRIEND
PZ0145 Sigma-Aldrich

PD-160725 2-hydroxyethanesulfonate

≥98% (HPLC)

Synonym: PD-160725 2-hydroxyethanesulfonic acid salt, PD-160725 isethionate salt

Purchase

Properties

Related Categories AMPA/Kainate Antagonists, Approved Therapeutics/Drug Candidates, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience,
InChI Key   PCVFALGXHRFVHN-UHFFFAOYSA-N
assay   ≥98% (HPLC)
form   powder, yellow
solubility   H2O: ≥20 mg/mL, clear, yellow
storage temp.   2-8°C

Description

Biochem/physiol Actions

PD-160725 2-hydroxyethanesulfonate is an AMPA receptor antagonist.

Features and Benefits

This compound was developed by Pfizer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. PZ0145.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Glutamate Receptors (Ion Channel Family)

The ion channel family of glutamate receptors ("€œionotropic"€ glutamate receptors or iGluR) comprises three major subtypes based on pharmacology and protein structure.
Keywords: Alzheimer Disease, Anticonvulsants, Atomic absorption spectroscopy, Clinical, Gene expression, Ligands, Neurotransmission, Physiological Processes, Schizophrenia

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?