EMAIL THIS PAGE TO A FRIEND
R2778 Sigma-Aldrich

RU 28362

≥98% (HPLC), solid

Synonym: (11β,17β)-11,17-Dihydroxy-6-methyl-17-(1-propynyl)-androsta-1,4,6-trien-3-one

Purchase

Properties

assay   ≥98% (HPLC)
form   solid
impurities   1.25 mol/mol Ethanol
solubility   DMSO: soluble 33 mg/mL
storage temp.   2-8°C
Gene Information   rat ... Nr3c1(24413)

Description

Biochem/physiol Actions

Glucocorticoid receptor agonist.

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?