• USA Home
  • R5755 - N-Acetyl-Renin Substrate Tetradecapeptide human

EMAIL THIS PAGE TO A FRIEND
R5755 Sigma-Aldrich

N-Acetyl-Renin Substrate Tetradecapeptide human

≥90% (HPLC)

Synonym: Ac-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Val-Ile-His-Asn

  • CAS Number 104180-27-0

  • Empirical Formula (Hill Notation) C85H124N24O20

  • Molecular Weight 1802.04

  •  MDL number MFCD00133569

Purchase

Properties

assay   ≥90% (HPLC)
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?