EMAIL THIS PAGE TO A FRIEND
S0399 Sigma-Aldrich

Sodium hydroxide

beads, 20-40 mesh

Synonym: ‘Caustic soda’

Purchase

Properties

vapor density   >1 (vs air)
vapor pressure   <18 mmHg ( 20 °C)
  3 mmHg ( 37 °C)
form   beads
particle size   20-40 mesh
mp   318 °C (lit.)
SMILES string   [OH-].[Na+]
InChI   1S/Na.H2O/h;1H2/q+1;/p-1
InChI key   HEMHJVSKTPXQMS-UHFFFAOYSA-M
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1823 8 / PGII
WGK Germany 
WGK 1
RTECS 
WB4900000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?