• USA Home
  • S1001 - Sodium phosphate tribasic dodecahydrate

EMAIL THIS PAGE TO A FRIEND
S1001 Sigma-Aldrich

Sodium phosphate tribasic dodecahydrate

≥98%

Synonym: TSP, Trisodium phosphate (tert) dodecahydrate, Trisodium phosphate dodecahydrate

Purchase

Properties

assay   ≥98%
density   1.62 g/mL at 25 °C
SMILES string   O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O
InChI   1S/3Na.H3O4P.12H2O/c;;;1-5(2,3)4;;;;;;;;;;;;/h;;;(H3,1,2,3,4);12*1H2/q3*+1;;;;;;;;;;;;;/p-3
InChI key   ASTWEMOBIXQPPV-UHFFFAOYSA-K
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
RTECS 
TC9575000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Buffers - From Bnech to Bulk
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

04277 Sodium phosphate tribasic dodecahydrate, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?