EMAIL THIS PAGE TO A FRIEND
S116 Supelco

(±)-Sulpiride

Synonym: (±)-5-(Aminosulfonyl)-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2-methoxybenzamide, (±)-N-1-(Ethylpyrrolidin-2-ylmethyl)-2-methoxy-5-sulfamoylbenzamide

Purchase

Properties

InChI Key   BGRJTUBHPOOWDU-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
BZ3400000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

S8010 (±)-Sulpiride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?