EMAIL THIS PAGE TO A FRIEND
S1320 Sigma-Aldrich

SR-202

≥98% (HPLC), crystalline

Synonym: Dimethyl (α-dimethoxyphosphinyl-p-chlorobenzyl) phosphate, Mifobate

  • CAS Number 76541-72-5

  • Empirical Formula (Hill Notation) C11H17ClO7P2

  • Molecular Weight 358.65

  •  MDL number MFCD00864813

  •  PubChem Substance ID 24724601

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Gene Regulation and Expression,
InChI Key   VQHUQHAPWMNBLP-UHFFFAOYSA-N
assay   ≥98% (HPLC)
form   crystalline
storage condition   desiccated
color   white
solubility   DMSO: 5 mg/mL
  H2O: insoluble
storage temp.   −20°C

Description

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

SR-202 is a selective PPARγ antagonist. Improves insulin sensitivity in diabetic mice.

Features and Benefits

This compound is featured on the Nuclear Receptors (PPARs) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Packaging

Product is air sensitive

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
TB8817000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?