• USA Home
  • S1417 - Saxitoxin diacetate salt

EMAIL THIS PAGE TO A FRIEND
S1417 Sigma-Aldrich

Saxitoxin diacetate salt

from Protogonyaulax sp., ≥70% (HPLC)

Synonym: STX

  • CAS Number 220355-66-8

  • Empirical Formula (Hill Notation) C10H17N7O4 · 2C2H4O2

  • Molecular Weight 419.39

  •  MDL number MFCD00134842

  •  NACRES NA.77

Purchase

Properties

biological source   Protogonyaulax sp.
assay   ≥70% (HPLC)
storage temp.   −20°C
SMILES string   CC(O)=O.CC(O)=O.NC(=O)OC[C@@H]1N=C(N)N2CCC(O)(O)[C@@]23NC(N)=NC13

Description

Biochem/physiol Actions

Selective Na+ channel blocker

Other Notes

Neurotoxin produced by dinoflagellates in "red tides."

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3462 6.1 / PGI
WGK Germany 
WGK 3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?