EMAIL THIS PAGE TO A FRIEND
S1648 Sigma-Aldrich

Sodium thiosulfate

anhydrous, ≥99%

Purchase

Properties

grade   anhydrous
assay   ≥99%
SMILES string   [Na+].[Na+].[O-]S([O-])(=O)=S
InChI   1S/2Na.H2O3S2/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2
InChI key   AKHNMLFCWUSKQB-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
RTECS 
XN6476000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?