• USA Home
  • S2136 - [Met-OH11]-Substance P

EMAIL THIS PAGE TO A FRIEND
S2136 Sigma-Aldrich

[Met-OH11]-Substance P

≥97% (HPLC)

  • CAS Number 71977-09-8

  • Empirical Formula (Hill Notation) C63H97N17O14S

  • Molecular Weight 1348.61

  •  MDL number MFCD00076781

  •  PubChem Substance ID 24899513

Purchase

Properties

Related Categories Agonist, Cell Biology, Cell Signaling and Neuroscience, Peptides and Proteins, Peptides for Cell Biology,
InChI Key   XHWDVRRNQHMAPE-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   human ... TAC1(6863)

Description

Amino Acid Sequence

Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met

Biochem/physiol Actions

NK-1 agonist

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?