EMAIL THIS PAGE TO A FRIEND
S4882 Sigma-Aldrich

Selenium sulfide

52-55.5%

Synonym: Selenium disulfide

Purchase

Properties

InChI Key   JNMWHTHYDQTDQZ-UHFFFAOYSA-N
assay   52-55.5%
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2657 6.1 / PGII
WGK Germany 
3
RTECS 
VS8925000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

257710 Selenium sulfide

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?