• USA Home
  • S5125 - L-Serine methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
S5125 Sigma-Aldrich

L-Serine methyl ester hydrochloride

  • CAS Number 5680-80-8

  • Linear Formula HOCH2CH(NH2)COOCH3 · HCl

  • Molecular Weight 155.58

  •  Beilstein/REAXYS Number 3559591

  •  EC Number 227-140-7

  •  MDL number MFCD00063680

Purchase

Properties

mp   163 °C (dec.) (lit.)
storage temp.   −20°C
SMILES string   Cl.COC(=O)[C@@H](N)CO
InChI   1S/C4H9NO3.ClH/c1-8-4(7)3(5)2-6;/h3,6H,2,5H2,1H3;1H/t3-;/m0./s1
InChI key   NDBQJIBNNUJNHA-DFWYDOINSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

412201 L-Serine methyl ester hydrochloride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?