• USA Home
  • S5382 - [p-Cl-Phe7,8]-Substance P

EMAIL THIS PAGE TO A FRIEND
S5382 Sigma-Aldrich

[p-Cl-Phe7,8]-Substance P

≥95% (HPLC)

  • CAS Number 73646-81-8

  • Empirical Formula (Hill Notation) C63H96Cl2N18O13S

  • Molecular Weight 1416.52

  •  MDL number MFCD00076786

  •  PubChem Substance ID 24899649

Purchase

Properties

assay   ≥95% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Arg-Pro-Lys-Pro-Gln-Gln-Phe(4-Cl)-Phe(4-Cl)-Gly-Leu-Met-NH2

Linkage

Substance P analog that can be tritiated

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?