• USA Home
  • S5515 - Sodium phosphate monobasic

EMAIL THIS PAGE TO A FRIEND
S5515 Sigma-Aldrich

Sodium phosphate monobasic

≥98%, BioReagent, suitable for plant cell culture

Synonym: Monosodium dihydrogen orthophosphate, Monosodium phosphate, Sodium dihydrogen phosphate

Purchase

Properties

InChI Key   AJPJDKMHJJGVTQ-UHFFFAOYSA-M
product line   BioReagent
assay   ≥98%
suitability   suitable for plant cell culture

Description

Application

Use in plant culture media pH adjustment.

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
WA1900000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?