• USA Home
  • S6314 - Sulforhodamine B acid chloride

EMAIL THIS PAGE TO A FRIEND
S6314 Sigma

Sulforhodamine B acid chloride

powder

Synonym: Lissamine rhodamine B sulfonylchloride

  • CAS Number 62796-29-6

  • Empirical Formula (Hill Notation) C27H29ClN2O6S2

  • Molecular Weight 577.11

  •  Beilstein Registry Number 7734927

  •  EC Number 263-735-8

  •  MDL number MFCD00042001

Purchase

Properties

InChI Key   YERWMQJEYUIJBO-UHFFFAOYSA-N
form   powder
color   brown to dark brown
fluorescence   λex 560 nm; λem 580 nm in chloroform
storage temp.   −20°C

Description

Application

Fluorescent protein label forming stable conjugates

Biochem/physiol Actions

Suitable for fluorescence labeling

Legal Information

Lissamine is a trademark of PerkinElmer Inc.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

86186 Sulforhodamine B acid chloride, suitable for fluorescence, technical

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?