• USA Home
  • S6393 - L-Serine-7-amido-4-methylcoumarin hydrochloride

EMAIL THIS PAGE TO A FRIEND
S6393 Sigma

L-Serine-7-amido-4-methylcoumarin hydrochloride

  • CAS Number 115918-60-0

  • Empirical Formula (Hill Notation) C13H14N2O4 · HCl

  • Molecular Weight 298.72

  •  MDL number MFCD00079565

  •  PubChem Substance ID 24899700

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Fluorogenic More...
InChI Key   RUMJTWWLPLESCZ-PPHPATTJSA-N
storage temp.   −20°C

Description

Packaging

25 mg in poly bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?