EMAIL THIS PAGE TO A FRIEND
S6640 Sigma-Aldrich

Swainsonine

from Rhizoctonia leguminicola

Synonym: (1S,2R,8R,8aR)-1,2,8-Octahydroindolizidinetriol

  • CAS Number 72741-87-8

  • Empirical Formula (Hill Notation) C8H15NO3

  • Molecular Weight 173.21

  •  Beilstein/REAXYS Number 4175740

  •  MDL number MFCD00017554

  •  PubChem Substance ID 329824566

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Inhibitors, Enzyme Inhibitors by Enzyme, Enzymes, Inhibitors, and Substrates, Glycobiology,
biological source   Rhizoctonia leguminicola
form   solid
storage temp.   −20°C
SMILES string   O[C@@H]1CCCN2C[C@@H](O)[C@@H](O)C12
InChI   1S/C8H15NO3/c10-5-2-1-3-9-4-6(11)8(12)7(5)9/h5-8,10-12H,1-4H2/t5-,6-,7?,8-/m1/s1
InChI key   FXUAIOOAOAVCGD-DCDLSZRSSA-N

Description

Application

Swainsonine is a potent inhibitor of various α-mannosidases, especially of α-mannosidase II. It inhibits glycoprotein processing and also acts as immune modulator.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
NM2408666

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

S8195 Swainsonine, from Metarrhizium anisopliae, ≥98% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?