EMAIL THIS PAGE TO A FRIEND
S6755 Sigma-Aldrich

Octadecylamine

~90%

Synonym: 1-Aminooctadecane, Stearylamine

  • CAS Number 124-30-1

  • Linear Formula CH3(CH2)17NH2

  • Molecular Weight 269.51

  •  Beilstein/REAXYS Number 636111

  •  EC Number 204-695-3

  •  MDL number MFCD00008159

Purchase

Properties

vapor pressure   10 mmHg ( 72 °C)
assay   ~90%
impurities   C16 and C17 homologs, balance
bp   232 °C/32 mmHg (lit.)
mp   50-52 °C (lit.)
SMILES string   CCCCCCCCCCCCCCCCCCN
InChI   1S/C18H39N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-19H2,1H3
InChI key   REYJJPSVUYRZGE-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Gastrointestinal tract, Immune system, Liver
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
RG4150000
Flash Point(F) 
298.4 °F
Flash Point(C) 
148 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

O1408 Octadecylamine, technical grade, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?