EMAIL THIS PAGE TO A FRIEND
S8518 Sigma-Aldrich

Sesamol

≥99% (TLC), crystalline

Synonym: 3,4-(Methylenedioxy)phenol, 5-Benzodioxolol

  • CAS Number 533-31-3

  • Empirical Formula (Hill Notation) C7H6O3

  • Molecular Weight 138.12

  •  Beilstein/REAXYS Number 127405

  •  EC Number 208-561-5

  •  MDL number MFCD00005827

  •  PubChem Substance ID 24899796

Purchase

Properties

assay   ≥99% (TLC)
form   crystalline
color   off-white to tan
mp   62-65 °C (lit.)
storage temp.   −20°C
SMILES string   Oc1ccc2OCOc2c1
InChI   1S/C7H6O3/c8-5-1-2-6-7(3-5)10-4-9-6/h1-3,8H,4H2
InChI key   LUSZGTFNYDARNI-UHFFFAOYSA-N

Description

Packaging

Packaged under Argon

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
SM0890000
Protocols & Articles

Related Content

Carcinogen Selector - Digestive System - Stomach

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Stomach
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Digestive System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System
Keywords: Cancer, Carcinogens, Respiratory

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?