• USA Home
  • S9389 - Spermidine-8-14C trihydrochloride

EMAIL THIS PAGE TO A FRIEND
S9389 Sigma-Aldrich

Spermidine-8-14C trihydrochloride

aqueous ethanol solution

  • CAS Number 133448-26-7

  • Linear Formula NH2[CH2]3NH(CH2)314(CH2)NH2 · 3HCl

  • Molecular Weight 254.63

  •  MDL number MFCD00213923

  •  PubChem Substance ID 24899838

Purchase

Properties

form   aqueous ethanol solution
extent of labeling   ≥10 mCi per mmol
storage temp.   2-8°C

Description

Packaging

serum bottle

Physical form

Aqueous solution containing 2% ethanol

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
-

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?