• USA Home
  • S9878 - N-Stearoyl-DL-dihydroglucocerebroside

EMAIL THIS PAGE TO A FRIEND
S9878 Sigma-Aldrich

N-Stearoyl-DL-dihydroglucocerebroside

Synonym: 1-O-(β-D-Glucopyranosyl)-N-octadecanoyl-DL-dihydrosphingosine

  • CAS Number 13032-63-8

  • Empirical Formula (Hill Notation) C42H83NO8

  • Molecular Weight 730.11

  •  MDL number MFCD00070178

Purchase

Properties

storage temp.   −20°C
SMILES string   CCCCCCCCCCCCCCCCCC(=O)NC(COC1OC(CO)C(O)C(O)C1O)C(O)CCCCCCCCCCCCCCC

Description

Linkage

Contains approx. 15% of the palmitoyl analog.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?