• USA Home
  • SAB1305581 - ANTI-PHOSPHO-IKKB(S675) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB1305581 Sigma

ANTI-PHOSPHO-IKKB(S675) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: I-kappa-B kinase 2, IKBKB, IKKB, Inhibitor of nuclear factor kappa-B kinase subunit beta, Nuclear factor NF-kappa-B inhibitor kinase beta

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PH-PI, Primary Antibodies
species reactivity   human
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   mol wt 86564 Da
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... IKBKB(3551)
biological source   rabbit
NCBI accession no.   NP_001177649.1
UniProt accession no.   O14920

Description

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Search & filter by validated application (ie, WB, IF/ICC, IHC, ELISA, etc,) host species, clonality (monoclonal, polyclonal and recombinant) and more. Learn from technical information, product highli...
Keywords: Amplification, Buffers, Enzyme-linked immunosorbent assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?