• USA Home
  • SAB1305585 - ANTI-PHOSPHO-ENT1(SLC29A1)(S254) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB1305585 Sigma

ANTI-PHOSPHO-ENT1(SLC29A1)(S254) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: ENT1, Equilibrative nitrobenzylmercaptopurine riboside-sensitive nucleoside transporter, Equilibrative nucleoside transporter 1, Nucleoside transporter, es-type, SLC29A1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PH-PI, Primary Antibodies
species reactivity   human
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   mol wt 50219 Da
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... SLC29A1(2030)
biological source   rabbit
NCBI accession no.   NP_001071642.1
UniProt accession no.   Q99808

Description

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Search & filter by validated application (ie, WB, IF/ICC, IHC, ELISA, etc,) host species, clonality (monoclonal, polyclonal and recombinant) and more. Learn from technical information, product highli...
Keywords: Amplification, Buffers, Enzyme-linked immunosorbent assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?