EMAIL THIS PAGE TO A FRIEND
SML1034 Sigma-Aldrich

MSOP

≥98% (HPLC)

Synonym: (R,S)-α-Methylserine-O-phosphate, M-SOP

  • CAS Number 66515-29-5

  • Empirical Formula (Hill Notation) C4H10NO6P

  • Molecular Weight 199.10

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Excitatory Amino Acids,
assay   ≥98% (HPLC)
form   powder
color   white to beige
solubility   H2O: 3 mg/mL, clear (warmed)
storage temp.   room temp

Description

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

MSOP, (RS)-α-Methylserine-O-phosphate, is a Group III metabotropic receptor antagonist with minimal activity on postsynaptic mGlu receptors or on ionotropic glutamate receptors.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis


Making Headway

Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Discover Bioactive Small Molecules for Neuroscience

The nervous system, comprised of the brain and spinal cord (central nervous system, or CNS) and network of nerves that connects the CNS to the rest of the body (peripheral nervous system, or PNS) is ...
Keywords: Bioactive small molecules, Central Nervous System, Diseases, Metabolism, Neurodegenerative Diseases, Neuroscience, Neurotransmission, Neurotransmitters

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?