EMAIL THIS PAGE TO A FRIEND
SML1588 Sigma-Aldrich

KDOAM25

≥97% (HPLC)

Synonym: 2-[[[2-[2-(Dimethylamino)ethyl-Ethyl-Amino]-2-Oxidanylidene-Ethyl]amino]methyl]pyridine-4-Carboxamide, 2-[[[2-[2-(dimethylamino)ethyl-ethylamino]-2-oxoethyl]amino]methyl]pyridine-4-carboxamide, GTPL8576, KDOAM-25, LQT

  • Empirical Formula (Hill Notation) C15H25N5O2

  • Molecular Weight 307.39

  •  NACRES NA.77

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, DNA-RNA Transcription Regulators,
Quality Level   100
assay   ≥97% (HPLC)
form   oil
color   white to beige
storage temp.   −20°C

Description

Biochem/physiol Actions

KDOAM25 is a selective inihbitor of the KDM5 family histone demethylases JARID1A, JARID1B, JARID1C and JARID1D with IC50 values < 60 nM. JARID1A and JARID1B are independently overexpressed in some cancers with JARID1B (KDM5B, PLU1) also identified as a potential oncogene, a repressor of tumour repressor genes. JARID1C (KDM5C) and JARID1D (KDM5D) are located on the X- and Y-chromosomes respectively. KDOAM25′s closest off-target is JMJD2C (selectivity >400 fold). It shows no activity on other tested 2-OG family members, including FIH, NO66, MINA53 and PHD2. KDOAM25 is active in cells.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?