EMAIL THIS PAGE TO A FRIEND
SRP6328 Sigma-Aldrich

CD300C human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym: CLM-6, CMRF-35, CMRF-35A, CMRF35-A1, CMRF35-like molecule 6, CMRF35, CMRF35A1, CMRF35A, IGSF16, LIR

  •  NACRES NA.32

Purchase

Properties

Related Categories CD160 to CD340, Cell Biology, Cell Signaling and Neuroscience, Cluster of Differentiation (CD) Proteins, Cytokines and Growth Factors,
biological source   human
recombinant   expressed in E. coli
assay   ≥85% (SDS-PAGE)
form   liquid
mol wt   20.2 kDa (186 aa, 21-183 aa + His tag)
packaging   pkg of 100 μg
concentration   1 mg/mL
impurities   <1 EU/μg endotoxin (LAL test)
UniProt accession no.   Q08708
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... CD300C(10871)

Description

General description

CMRF35-like molecule 6, also known as CD300C, belongs to the CD300 family. The CMRF35 antigen, which was identified by reactivity with a monoclonal antibody, is present on monocytes, neutrophils, and some T and B lymphocytes. Recombinant human CD300C protein, fused to His-tag at N-terminus, was expressed in E.coli.

Physical form

1 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.4 M Urea and 10% glycerol.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?